Notice: This product will be discontinued on May1, 2017. Please contact Customer Service for possible alternatives.

Optimized DyNAzyme Buffer (10X)

Catalog number: F511L

Thermo Scientific™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Optimized Thermo Scientific 10X DyNAzyme buffers are available in several formats: with and without detergents as well as in Mg2+-free solution.
Includes

Optimized 10X DyNAzyme Buffer (F-511):

• 1X buffer contains 10 mM Tris-HCl (pH 8.8 at 25°C), 1.5 mM MgCl2, 50 mM KCl, and 0.1% Triton X-100
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: DyNAzyme II DNA Polymerase
Product Line: DyNAzyme
Product Size: 4 x 1.5 mL

Contents & storage

Contains:
• 4 x 1.5 mL 10X DyNAzyme Buffer Store at -20°C.

Documents

Manuals & protocols