Search Thermo Fisher Scientific
Thermo Scientific™ FHT 59 N1 Nuclide-Specific Aerosol Monitor continuously monitors aerosol-bound gamma activity concentration in air or effluent gas. It is nuclide-specific detection of gamma-emitting isotopes.