Yeast Nitrogen Base, without amino acids, without ammonium sulfate
Yeast Nitrogen Base, without amino acids, without ammonium sulfate
Thermo Scientific Chemicals
Yeast Nitrogen Base, without amino acids, without ammonium sulfate
Used for classifying yeasts based on carbohydrate and amino acids requirements. Recommended for use in classifying yeasts based on carbon and nitrogen requirements. Contains all essential nutrients required for the growth of yeasts except amino acids, nitrogen and carbohydrate.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number H26271.36
also known as H26271-36
Price (USD)
630.65
Online Exclusive
701.00
Save 70.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
630.65
Online Exclusive
701.00
Save 70.35 (10%)
Each
Add to cart
Specifications
Packaging TypePlastic Bottle
Quantity500 g
Recommended StorageAmbient temperatures
FormPowder
Unit SizeEach
Yeast Nitrogen Base without Amino Acids and Ammonium Sulfate is used for classifying yeasts based on carbohydrate and amino acids requirements. Recommended for use in classifying yeasts based on carbon and nitrogen requirements. Contains all essential nutrients required for the growth of yeasts except amino acids, nitrogen and carbohydrate. Addition of nitrogen and carbon sources is required.