Easily visualize entire cells or individual cell structures during high-content screening (HCS) assays with HCS CellMask stains, which are available in a variety of colors for multiplexing flexibility and can be used immediately after fixation and permeabilization, or after antibody labeling.
Have Questions?
보기 방식 변경
카탈로그 번호
설명
H32714
HCS CellMask Green Stain
H32720
HCS CellMask Blue Stain
H32722
HCS CellMask Near-IR Stain
H32713
HCS CellMask Orange Stain
H32712
HCS CellMask™ Red Stain
H32721
HCS CellMask Deep Red Stain
6 개 옵션
카탈로그 번호 H32714
제품 가격(KRW)
591,000
온라인 행사
Ends: 31-Dec-2026
695,000
할인액 104,000 (15%)
Each
설명:
HCS CellMask Green Stain
제품 가격(KRW)
591,000
온라인 행사
Ends: 31-Dec-2026
695,000
할인액 104,000 (15%)
Each
Identify entire cells (cytoplasm and nucleus) during high-content screening (HCS) assays with the versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains, which are fluorophores that can be used immediately after fixation and permeabilization, or after antibody labeling. They are available in a variety of colors for multiplexing protocols.
The versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains are cell delineation tools for high content screening (HCS) platforms, label the entire cell (cytoplasm and nucleus) and provide an accurate backdrop against which the features of interest can be assessed. The HCS CellMask Red and Blue stains replace the HCS CellMask Red cytoplasmic/nuclear (Cat. No. H32711) and HCS CellMask Blue cytoplasmic/nuclear stains (Cat. No. H34558), respectively.
HCS CellMask stains can be applied to cells immediately after fixation or in the last step of multiplexing protocols, and they are compatible with detergent-based cell permeabilization protocols. If only nuclear staining is desired, HCS NuclearMask stains can be used for measuring DNA content in addition to enabling robust cell demarcation. HCS NuclearMask stains are also available in a choice of colors for multiplexing flexibility.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
사양
색상Green
설명HCS CellMask Green Stain
검출 방법Fluorescence
방출Visible
여기 파장 범위493/516
용도(장비)High Content Instrument
제품라인CellMask
수량1 Set
배송 조건Room Temperature
라벨 유형Fluorescent Dye
제품 유형Stain
Sub Cellular LocalizationCytoplasm, Cytosol, Nucleus
Unit SizeEach
구성 및 보관
Includes 1 set of vials, providing sufficient material to stain a total of ten 96-well microplates.
Store at ≤-20°C, desiccate, and protect from light.
자주 묻는 질문(FAQ)
What dyes are used to make the CellMask stains?
The proprietary fluorescent dyes in the CellMask stains are general cytoplasmic stains. They are not found to bind to any specific cellular component.
Brown fat determination and development from muscle precursor cells by novel action of bone morphogenetic protein 6.
Authors:Sharma A, Huard C, Vernochet C, Ziemek D, Knowlton KM, Tyminski E, Paradis T, Zhang Y, Jones JE, von Schack D, Brown CT, Milos PM, Coyle AJ, Tremblay F, Martinez RV,
Journal:
PubMed ID:24658703
Brown adipose tissue (BAT) plays a pivotal role in promoting energy expenditure by the virtue of uncoupling protein-1 (UCP-1) that differentiates BAT from its energy storing white adipose tissue (WAT) counterpart. The clinical implication of ... More
Approaches to protozoan drug discovery: phenotypic screening.
Authors:Sykes ML, Avery VM,
Journal:
PubMed ID:23927763
Determining the activity of a compound and the potential impact on a diseased state is frequently undertaken using phenotypic or target-based approaches. Phenotypic screens have the advantage of the whole organism being exposed to the compound and thus all the targets and biological pathways associated with it. Cell penetration and ... More
Effect of microgrooved surface topography on osteoblast maturation and protein adsorption.
Authors:De Luca AC, Zink M, Weidt A, Mayr SG, Markaki AE,
Journal:
PubMed ID:25630566
Microgrooved surfaces have been used extensively to influence cell contact guidance. Guiding cell growth, extracellular matrix deposition, and mineralization is important for bone implant longevity. In this study, we investigated the osteoblast response to microgrooved metallic surfaces in serum-supplemented medium. Groove spacing was comparable with the spread osteoblast size. Focal ... More
BMP signaling in astrocytes downregulates EGFR to modulate survival and maturation.
Astrocytes constitute a major cell population in the brain with a myriad of essential functions, yet we know remarkably little about the signaling pathways and mechanisms that direct astrocyte maturation. To explore the signals regulating astrocyte development, we prospectively purified and cultured immature postnatal rodent astrocytes. We identified fibroblast growth ... More
Simultaneous flow cytometric analysis of IFN-? and CD4 mRNA and protein expression kinetics in human peripheral blood mononuclear cells during activation.
Authors:Van Hoof D, Lomas W, Hanley MB, Park E,
Journal:
PubMed ID:25123411
The application of fluorescently-labeled antibodies for flow cytometric identification and characterization of specific cell types within heterogeneous populations by their protein expression profile is well established. While detection of proteins is informative, concomitant transcript analysis in the same cells would provide a more complete and comprehensive view of intracellular signaling ... More