It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
M. S. Frasinyuk; V. P. Khilya. Preparation and reactions of isoflavone heteroanalogs. Chemistry of Heterocyclic Compounds,. 1999, 35(1), 3-22.
O. V. Khilya; M. S. Frasinyuk; A. V. Turov; V. P. Khilya. Chemistry of 3-Hetarylcoumarins. 1. 3-(2-Benzazolyl)coumarins. Chemistry of Heterocyclic Compounds., 2001, 37(8), 1029-1037.