Thermo Scientific Chemicals

(4-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: H55534.06
5 g, Each
Thermo Scientific Chemicals

(4-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals

Catalog number: H55534.06
5 g, Each
Quantity
Catalog number: H55534.06
also known as H55534-06
Price (USD)
Price: 107.00
Online price: 91.65
Your price:
Quantity
-

Chemical Identifiers

CAS
16116-78-2
IUPAC Name
[2-(4-bromophenyl)ethynyl]trimethylsilane
Molecular Formula
C11H13BrSi
InChI Key
RNMSGCJGNJYDNS-UHFFFAOYSA-N
SMILES
C[Si](C)(C)C#CC1=CC=C(Br)C=C1
Assay (GC)
>97.5%

Description

It is used as the intermediate of Organic Light Emitting Diode (OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as the intermediate of Organic Light Emitting Diode (OLED).

Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text