Search Thermo Fisher Scientific
Catalog Number | Detectors | Description |
---|---|---|
HFM11A-GF-P1R | 7: 4 Hands, 2 Feet, 1 frisking probe | Gas flow detect counters 4 hand, 2 foot, frisker housing |
HFM11A-GF | 6: 4 Hands, 2 Feet | Gas flow detector counters 4 hand, 2 foot |
HFM11A-SC | 6: 4 Hands, 2 Feet | Alpha Beta scintillation (4 hand and 2 foot detectors) |
HFM11A-SC-P3R | 7: 4 Hands, 2 Feet, 1 frisking probe | Alpha Beta scintillation (fisker housing detectors) |
Thermo Scientific™ HFM11 Series high-efficiency Hand and Foot Monitors help to reduce monitoring times; the longer hand detectors provide wrist and forearm coverage and there is an optional frisker probe available for additional monitoring.
The HFM11 provides fast, thorough, hand and foot monitoring with class-leading reliability and ease of use.
Includes:Sophisticated voltage scanning software