Invitrogen™
Image-iT™ Cell Painting Kit
Catalog number: I63000
Invitrogen™
Image-iT™ Cell Painting Kit
Catalog number: I63000
Catalog Number
I63000
Unit Size
1 kit
Price (USD)
1,110.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
I630001 kit
1,110.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Image-iT Cell Painting Kit is a collection of reagents that can be used to check cell health/cytotoxicity by “painting” different cellular components. When combined with a high-content imager, such as the CellInsight platform, these reagents can provide a phenotypical readout of cell status. This kit is based on the publication by Bray et al. (1) describing the morphological profiling of cells using multi-plexed fluorescent dyes.
This kit includes the following reagents (1 each):
• Hoechst 33342, Trihydrochloride, Trihydrate (Cat. No. H21492)
• Concanavalin A, Alexa Fluor 488 Conjugate (Cat. No. C11252)
• Wheat Germ Agglutinin, Alexa Fluor 555 Conjugate (Cat. No. W32464)
• Alexa Fluor 568 Phalloidin (Cat. No. A12380)
• SYTO RNASelect Green Fluorescent Cell Stain (Cat. No. S32703)
• MitoTracker Deep Red FM (Cat. No. M22426)
Reference
1. Bray MA, Singh S, Han H, Davis CT, Borgeson B, Hartland C, Kost-Alimova M, Gustafsdottir SM, Gibson CC, Carpenter AE. Cell Painting, a high-content image-based assay for morphological profiling using multiplexed fluorescent dyes. Nat Protoc. 2016 Sep;11(9):1757-74.