Nanospray Ion Source
Thermo Scientific™
Nanospray Ion Source
Increase the sensitivity of LC-MS analyses with the Thermo Scientific™ Nanospray ion source.
Have Questions?
Catalog NumberType
IQLAAEGABBFADNMAIFNanospray Ion Source
Catalog number IQLAAEGABBFADNMAIF
Price (USD)
-
Type:
Nanospray Ion Source
Increase the sensitivity of LC-MS analyses with nanospray ionization. The Thermo Scientific™ Nanospray ion source is a comprehensive solution with a high-resolution imaging system for direct observation and optimization of the spray. Each nanospray source is supplied with a consumables kit, allowing quick startup.
Complete low-flow solution with source, imaging, consumables, instructions, and support
  • Easily interchangeable probe design supports static, dynamic, and packed-tip nanospray
  • Accommodates integrally packed fused-silica needle tip for zero post-column analyte loss
  • High-voltage connection enables application of voltage to the tip or to a liquid junction without repositioning the probe, reducing setup time
  • Picture-in-picture allows spray visualization on the computer monitor for ease of tuning and positioning
Specifications, terms, and pricing are subject to change. Not all products are available in all countries. Please consult your local sales representative for details.
Specifications
TypeNanospray Ion Source
Unit SizeEach