Prelude SPLC™ System
Thermo Scientific™  Related applications: Industrial Mass Spectrometry
Analyze complex biological samples quickly, reproducibly and reliably using the Thermo Scientific™ Prelude SPLC™ System with multiplexing and Thermo Scientific™ TurboFlow™ Technology. Combined with a mass spectrometer from Thermo Scientific, the Prelude SPLC System speeds and simplifies on-line clean up, high-performance chromatographic separation and mass spectrometric analysis of complex biological samples. The selectivity, versatility and quantitative accuracy of LC/MS are now accessible and economically viable to the clinical research and forensic toxicology lab.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
Back to top
Description
Multiplexing and TurboFlow Technology
  • Chromatographic multiplexing maximizes mass spectrometer use, sample throughput, and assay flexibility 
  • Online sample clean up using TurboFlow technology saves time, eliminates matrix interferences and ion suppression, and reduces variably 
  • Designed for use with ultra high efficiency small particle size columns 
Easy to Use
  • Preprogrammed application-specific methods speed setup 
  • Easy, error-free, tool-less consumable replacement
  • On-site, method-specific training included
Maximizes Uptime, Minimizes Costs
  • Designed for reliability and ease of service to reduce maintenance costs
  • Simplified operation means an expert operator is not required
  • TurboFlow technology reduces reagent costs
  • High-efficiency methods reduce solvent consumption
  • Optional remote diagnostics ensure rapid resolution of any problems that arise
For research use only. Not for use in diagnostic procedures. Availability may vary by country.
New Clinical Research and Forensic Toxicology Application Compendium
Discover the state of the art in LC-MS analysis for clinical research, drug monitoring research, forensic toxicology, sports anti-doping, and translational research.
Download the compendium
Documents
Manuals & protocols