Description
Ethylenediaminetetraacetic acid solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, chelators and ethylenediaminetetraacetic acid (EDTA) and EDTA solutions. Ethylenediaminetetraacetic acid solution has been used in a study to help elucidate the role of antigen retrieval in immunostaining of ethanol-fixed smears. Ethylenediaminetetraacetic acid solution has also been used in a study to assess the uptake and release of biotin-labeled fluorescein (b-FITC) as well as immobilization of biotin-labeled glucose oxidase (b-GOx) to the polyelectrolyte microcapsules with an avidin-poly(styrene sulfonate) (PSS) membrane.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Ethylenediaminetetraacetic acid solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, chelators and ethylenediaminetetraacetic acid (EDTA) and EDTA solutions. Ethylenediaminetetraacetic acid solution has been used in a study to help elucidate the role of antigen retrieval in immunostaining of ethanol-fixed smears. Ethylenediaminetetraacetic acid solution has also been used in a study to assess the uptake and release of biotin-labeled fluorescein (b-FITC) as well as immobilization of biotin-labeled glucose oxidase (b-GOx) to the polyelectrolyte microcapsules with an avidin-poly(styrene sulfonate) (PSS) membrane.
Solubility
Soluble in water.
Notes
Not tested for endtoxin or mycoplasma contaminants
RUO – Research Use Only