Thermo Scientific Chemicals

Ready-Made LB Agar Powder, Thermo Scientific Chemicals

Catalog number: J75851.A1
1 kg, Each
Thermo Scientific Chemicals

Ready-Made LB Agar Powder, Thermo Scientific Chemicals

Catalog number: J75851.A1
1 kg, Each
Quantity

Description

pH(3.5%, H2O): 7.0 ± 0.2
Contents:
  • Casein Peptone: 10gm/L
  • Yeast Extract: 5gm/L
  • Sodium Chloride: 5gm/L
  • Agar: 15gm/L
RUO – Research Use Only