Product Image
Invitrogen™

GeneBLAzer™ GIPR-CRE-bla HEK 293T Cells

GeneBLAzer® GIPR-CRE-bla HEK 293T cells contain the human Gastric Inhibitory Peptide Receptor (GIPR), stably integrated into the CellSensor® CRE-bla HEKRead more
Have Questions?
Catalog NumberQuantity
K18641 vial
Catalog number K1864
Price (USD)
11,840.00
Each
Quantity:
1 vial
Price (USD)
11,840.00
Each
GeneBLAzer® GIPR-CRE-bla HEK 293T cells contain the human Gastric Inhibitory Peptide Receptor (GIPR), stably integrated into the CellSensor® CRE-bla HEK 293T cell line. CellSensor® CRE-bla HEK 293T cells (Cat. no. K1535)) contain a beta-lactamase reporter gene under control of the CRE Response Element. The GeneBLAzer® GIPR-CRE-bla HEK 293T cells are functionally validated for Z’-factor and EC50 concentrations of Gastric Inhibitory Peptide (GIP). In addition, GeneBLAzer® GIPR-CRE-bla HEK 293T cells have been tested for assay performance under variable conditions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_000164.2
Assay EntryCell-based beta lactamase reporter gene
Cell StateDividing Cells
For Use With (Application)FRET
Human Protein ClassG-Protein Coupled Receptor (GPCR)
Population StatusSingle Cell Clone Origin
Product LineGeneBLAzer
Quantity1 vial
ReadoutEnd Point
Target EntryGIPR
Cell Line293T (HEK)
Detection MethodFluorescence
Druggable TargetGPCR
System TypeGeneBLAzer™
TypeCell Line
Unit SizeEach
Contents & Storage
GeneBLAzer™ GIPR-CRE-bla HEK 293T cells are shipped on dry ice, 8 x 10^6 cells⁄vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.