PureLink™ HiPure Plasmid Filter Maxiprep Kit

Catalog number:  K210017

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PureLink® HiPure Plasmid Filter Maxiprep Kit is designed to isolate plasmid DNA of the highest purity from 100 to 500 mL of an overnight bacterial culture. The PureLink® HiPure Plasmid Filter Kits include HiPure Filter Columns, which combine filtration for clarification of the bacterial lysate with anion-exchange resin purification of plasmid DNA, in a single unit. The resulting plasmid DNA is of a quality equivalent to that obtained by purification with two passes through cesium chloride gradients—the most rigorous method for DNA purification. Purify high-quality DNA in less than two hours for transfections and other applications with no need for additional steps to remove contaminants like RNA, proteins, or endotoxins. Additionally, the elimination of phenol, chloroform, ethidium bromide, and cesium chloride from the protocol minimizes exposure to, and disposal of hazardous materials. The PureLink® HiPure Plasmid Filter Kits provide:

Simple protocols—bacterial lysate clarification without centrifugation
Fast results—save time by purifying plasmid DNA in less than two hours
High yields—up to 200 µg for midipreps and up to 850 µg for maxipreps (see table)
Reliable performance—the high-quality plasmid DNA purified with these kits is suitable for a wide variety of applications

How it works
The addition of a HiPure Filter Column to the PureLink® HiPure Plasmid Kits provides rapid clearing of the bacterial lysate without the need for centrifugation (see figure). The lysate filtration cartridge is integrated into the DNA-binding column for a combined step of lysate clarification and plasmid DNA binding directly to the anion-exchange resin. A single column wash removes impurities such as RNA, proteins, metabolites, and other low molecular weight molecules; the ultrapure plasmid DNA is then eluted with a high-salt buffer. The DNA is then desalted and concentrated with an isopropanol precipitation step, then collected by centrifugation. The entire protocol can be completed in about two hours.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

LoyaltyEligible: SKUEligibleForLoyalty
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Sample Type (General): Bacteria
Downstream Application: Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Elution Volume - max: 15 ml
Elution Volume - min: 15 ml
Final Product: Plasmid DNA
Isolation Technology: Anion Exchange Resin with Lysate Clearing Filter
Number of Reactions: 25 Preps
Plasmid Type: <40kb, BAC, High Copy Plasmid, Low Copy Plasmid
Product Line: PureLink™
Product Size: 25 preps
Purification Time: 90 min
Purity or Quality Grade: Transfection Grade
Quantity: 25 columns
Sample Type (Specific): Bacterial Culture
System: PureLink™
Yield (up to): 1000 µg
Shipping Condition: Room Temperature

Contents & storage

The PureLink® HiPure Plasmid Filter Kits include Equilibration Buffer (EQ1), Resuspension Buffer (R3), Lysis Buffer (L7), Precipitation Buffer (N3), RNase A, Wash Buffer (W8), Elution Buffer (E4), TE Buffer (TE), and HiPure Filter Columns. Store all components at room temperature upon receipt.

Documents

Manuals & protocols

Recommended products