PureLink® Pro 96 Buffer Kit

Catalog number: K211000

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

PureLink® Pro 96 Buffer Kit is a convenient buffer kit enabling to separately purchase all buffers needed for the PureLink® Pro Plasmid Purification Kit (K2110-04A and K2110-24A). The buffer set comes with five buffers (resuspension-, lysis-, neutralization-, and wash buffers). The volume is 600 ml each and the wash buffer has a volume of 864 ml. The total buffer volume is >3L. In addition, RNase A is provided as 24 ml (20 mg⁄ml) and two empty bottles for making dilutions are provided.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): Bacteria
High Throughput Compatibility: High Throughput-Compatible
Downstream Application: Cloning, Next-Generation Sequencing, PCR, Sequencing
Final Product: Plasmid DNA
Isolation Technology: Silica Plate
Product Line: PureLink™
Product Size: 1 kit
Sample Type (Specific): Bacterial Culture
Starting Material (Volume): 1-3 ml
Yield (up to): 800 µg
Shipping Condition: Room Temperature

Contents & storage

Set of 6 components of which 5 are buffers and one is RNase A. Store all components at room temperature

Documents

Manuals & protocols