Zero Blunt&trade; TOPO&trade; PCR Cloning Kit, with One Shot&trade; TOP10 Chemically Competent <i>E. coli</i> (Supply Center Packaging)
Invitrogen™

Zero Blunt™ TOPO™ PCR Cloning Kit, with One Shot™ TOP10 Chemically Competent E. coli (Supply Center Packaging)

Zero Blunt™ TOPO™ PCR Cloning Kit provides a highly efficient, 5-minute, one-step cloning strategy ("TOPO™ Cloning") for the direct insertionRead more
Have Questions?
Catalog NumberQuantity
K280020SC25 Reactions
Catalog number K280020SC
Price (USD)
883.65
Online exclusive
940.00
Save 56.35
Each
Quantity:
25 Reactions
Price (USD)
883.65
Online exclusive
940.00
Save 56.35
Each
Zero Blunt™ TOPO™ PCR Cloning Kit provides a highly efficient, 5-minute, one-step cloning strategy ("TOPO™ Cloning") for the direct insertion of blunt-ended PCR products amplified with proofreading thermostable polymerases into a plasmid vector. Each kit has the Zero Blunt™ TOPO™ vector (see figure) containing the ccdB gene for positive selection, only permitting growth of plasmid vectors with recombinants. Zero™ Blunt™ TOPO™ kits are available with a variety of competent cells, or without competent cells, depending on your needs and budget. This product comes in Supply Center packaging, which features an additional outer box that holds both the TOPO™ vector box and the competent cells box. This special packaging helps ensure that visitors to a Supply Center obtain the complete kit.

Features of the Zero™ Blunt™ TOPO™ PCR Cloning Kits:

Fast and easy—go from PCR to clones in just 3 steps and in as little as 5 minutes hands-on time
Efficient—obtain up to 95% clones with correct insert
Proven—reliable performance for over a decade with over 4,000 citations
Simple —no ligase, post-PCR procedures, or PCR primers containing specific sequences are required.

Zero Blunt™ TOPO™ Cloning Kits—overview

Vector: pCR™Blunt II-TOPO™ vector—contains the ccdB gene, allowing for direct selection of recombinants

Cloning method: TOPO™ cloning—Topoisomerase I–based, 5-minute ligation of blunt-ended PCR products amplified with proofreading thermostable polymerases

Competent cells: Various options—choose from kits with either general, high-efficiency, bacteriophage T1-resistant, fast-growing competent cells, or use your own.

pCR™Blunt II-TOPO™ vector—designed for proofreading polymerases
The pCR™Blunt II-TOPO™ vector is designed to clone blunt-ended PCR products generated by thermostable proofreading polymerases such as Platinum™ Pfx DNA Polymerase. The pCR™Blunt II-TOPO™ vector contains:

• EcoRI sites flanking the PCR product insertion site for easy excision of inserts
• Kanamycin and Zeocin™ resistance genes for your choice of selection in E. coli
• SP6 promoter/primer site for in vitro RNA transcription and sequencing
• M13 forward and reverse primer sites for sequencing or PCR screening

pCR™Blunt II-TOPO™ clone selection
pCR™-Blunt II-TOPO™ allows for direct selection of recombinants via disruption of the lethal E. coli gene, ccdB. The vector contains the ccdB gene fused to the C-terminus of the LacZα fragment. Ligation of a blunt-end PCR product disrupts expression of the LacZα-ccdB gene fusion, permitting growth of only positive recombinants upon transformation. Cells that contain non-recombinant vector are killed upon plating. Therefore, blue/white screening is not required.

Simplified TOPO™-based cloning
Using TOPO™ cloning technology, there is no need for PCR primers containing specific sequences, post-PCR procedures, vector preparation, or other time-intensive DNA manipulation steps. Just add your PCR reaction straight to the provided topoisomerase-charged vector, incubate 5 minutes, and transform E. coli competent cells.

Efficient cloning
With up to 95% of clones carrying the desired insert, you can screen less clones, saving you time and money. The pCR™Blunt II-TOPO™ vector used in this kit comes with lethal ccdB gene for efficient selection of blunt-ended PCR recombinants products generated bythermostable proofreading polymerases.

The standard in cloning
When it comes to cloning, TOPO™ cloning technology has been a reliable partner for thousands of scientists for over ten years. Fast, simple-to-use, and efficient, TOPO™ cloning has been applied to many different vectors for a wide array of applications.

Zero Blunt™ TOPO™ PCR Cloning™ Kit options
The Zero Blunt™ TOPO™ PCR Cloning Kit for direct insertion of blunt-ended PCR products into a plasmid vector can be purchased with a variety of competent cells that deliver different advantages depending upon your needs:

General cloning: TOP10 cells (Cat. No. K2800-20, K2800-40)
High-efficiency cloning: TOP10 Electrocomp™ Cells (Cat. No. K2860-01, K2860-40)
General cloning, bacteriophage T1 resistance: DH5α-T1R (Cat. No. K2820-20, K2820-40)
Fast growth: Mach1™-T1R Chemically Competent E. Coli (Cat. No. K2830-20)
Provide your own: for flexibility and to save money (Cat. No. 450254)

We also offer a version of the kit that includes a PureLink™ Quick Plasmid Miniprep Kit (Cat. No. K2800-02) for use in isolation of clean, sequencing-ready, recombinant plasmid.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bacterial or Yeast StrainTOP10
Cell TypeChemically Competent
Cloning MethodBlunt TOPO™
Product LineOne Shot
Product TypePCR Cloning Kit
PromoterSP6
Quantity25 Reactions
Shipping ConditionDry Ice
VectorBlunt DNA Cloning Vectors
FormatKit
Unit SizeEach
Contents & Storage
Box 1:
• Linearized and topoisomerase I-activated pCR™Blunt II-TOPO™ vector
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid

Store at -68 to -85°C.

Frequently asked questions (FAQs)

What is the difference between the pCR2.1-TOPO and pCR4-TOPO vectors?

The vector backbones for both of these vectors are very similar. The main difference is that the pCR4-TOPO vector has sequencing primer sites located as close as 33 base pairs from the PCR product insertion site. This minimizes the amount of vector DNA sequence that needs to be read before reaching the sequence of the insert, making the pCR4-TOPO vector very useful for sequencing applications.

What is the difference between the pCR2.1-TOPO and pCRII-TOPO vectors?

The vector backbones for both of these vectors are very similar. The main difference is that the pCRII-TOPO vector is a dual promoter vector, containing the SP6 and T7 promoters for in vitro transcription/sequencing, whereas the pCR2.1-TOPO vector contains only the T7 promoter for in vitro transcription/sequencing. Both vectors contain the M13 Forward and Reverse primer sites for sequencing or PCR screening.

Your TOPO cloning kits contain a control template and control primers. Can I obtain the sequence of the control template?

The sequence of the control template is proprietary.

What is the best molar ratio of PCR product:vector to use for TOPO TA cloning? Is there an equation to calculate the quantity to use?

We suggest starting with a molar ratio of 1:1 (insert:vector), with a range of 0.5:1 to 2:1. The quantity used in a TOPO cloning reaction is typically 5-10 ng of a 2 kb PCR product.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 (insert:vector ratio)

What is the best ratio of insert:vector to use for cloning? Is there an equation to calculate this?

The optimal ratio is 1:1 insert to vector. Optimization can be done using a ratio of 0.5-2 molecules of insert for every molecule of the vector.

Equation:

length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 insert:vector ratio

Citations & References (3)

Citations & References
Abstract
Contribution of acquired carbapenem-hydrolyzing oxacillinases to carbapenem resistance in Acinetobacter baumannii.
Authors:Héritier C, Poirel L, Lambert T, Nordmann P
Journal:Antimicrob Agents Chemother
PubMed ID:16048925
'Carbapenem-hydrolyzing oxacillinases are reported increasingly in Acinetobacter baumannii. Since they hydrolyze carbapenems at low levels, the roles of carbapenem-hydrolyzing oxacillinases OXA-23, OXA-40, and OXA-58 in A. baumannii were determined. The bla(OXA-23), bla(OXA-40), and bla(OXA-58) genes were inserted in broad-host-range plasmid pAT801 and transformed in Escherichia coli DH10B and in A. ... More
A gene network for long-day flowering activates RFT1 encoding a mobile flowering signal in rice.
Authors:Komiya R, Yokoi S, Shimamoto K
Journal:Development
PubMed ID:19762423
'Although some genes that encode sensory or regulatory elements for photoperiodic flowering are conserved between the long-day (LD) plant Arabidopsis thaliana and the short-day (SD) plant rice, the gene networks that control rice flowering, and particularly flowering under LD conditions, are not well understood. We show here that RICE FLOWERING ... More
Transmission of human immunodeficiency virus type 1 from a patient who developed AIDS to an elite suppressor.
Authors:Bailey JR, O'Connell K, Yang HC, Han Y, Xu J, Jilek B, Williams TM, Ray SC, Siliciano RF, Blankson JN
Journal:J Virol
PubMed ID:18495769
Elite suppressors (ES) are untreated human immunodeficiency virus type 1 (HIV-1)-infected patients who maintain viral loads of <50 copies/ml. The mechanisms involved in this control of viral replication remain unclear. Prior studies suggested that these patients, as well as long-term nonprogressors, are infected with defective HIV-1 variants. Other reports have ... More