Product Image
Invitrogen™
DES™-Blasticidin Support Kit
DES™-Blasticidin Support Kitは、あらゆるDES™発現ベクターと組み合わせて使用できるように設計されており、ブラストサイジンを使用して安定したS2細胞を迅速に選択できます。DES™-Blasticidin Support Kitには、ブラストサイジンを使用して安定したS2細胞株を迅速に生成するためのpCoBlast選択ベクターが含まれています。このキットをpMT/V5-His詳細を見る
製品番号(カタログ番号)数量
K515001
または、製品番号K5150-01
1 kit
製品番号(カタログ番号) K515001
または、製品番号K5150-01
価格(JPY)
212,500
Each
数量:
1 kit
DES™-Blasticidin Support Kitは、あらゆるDES™発現ベクターと組み合わせて使用できるように設計されており、ブラストサイジンを使用して安定したS2細胞を迅速に選択できます。DES™-Blasticidin Support Kitには、ブラストサイジンを使用して安定したS2細胞株を迅速に生成するためのpCoBlast選択ベクターが含まれています。このキットをpMT/V5-His、pMT/V5-His-TOPO™、pMT-DES48、pMT/BIP/V5-His、または pAc5.1/V5-His、ベクターおよびDES™-BlasticidIn Support Kitと組み合わせることで、ショウジョウバエS2細胞での安定した発現を実現するa完全なシステムが形成されます。
研究用にのみ使用できます。診断用には使用いただけません。
仕様
製品タイプBlasticidin Support Kit
数量1 kit
ベクターpCoBlast
製品ラインDES
Unit SizeEach
組成および保存条件
DES™-Blasticidin Support Kitには、20 µgのpCoBlast、Calcium Phosphate Transfection Kit、凍結されたS2細胞、GIBCO™ Schneiderのショウジョウバエ培地、および50 mgのブラストサイジンが含まれます。S2細胞を液体窒素に保管してください。培地は+4℃で保管してください。その他のすべての試薬を-20℃で保管してください。すべての構成品は、適切に保存した場合、6カ月間保証されます。
よくあるご質問(FAQ)
Do I need to include a Kozak sequence for expression of recombinant proteins in insect cells?
While the importance of a Kozak consensus sequence in translation initiation has been demonstrated in mammalian cells, there seems to be some debate as to whether the Kozak rules are as stringent in insect cells. The only way to determine its importance would be a direct comparison of expression of the same protein from different initiation sequences. Even then, the rules for optimal expression of one protein may not hold for another. Here are two references which indicate that a Kozak consensus sequence does not have any effect on efficiency of expression in insect cells:
- Hills D, Crane-Robinson C (1995) Baculovirus expression of human basic fibroblast growth factor from a synthetic gene: role of the Kozak consensus and comparison with bacterial expression.
- Biochim Biophys Acta 1260(1):14-20.
- Ranjan A, Hasnain SE (1995) Influence of codon usage and translational initiation codon context in the AcNPV-based expression system: computer analysis using homologous and heterologous genes. Virus Genes 9(2):149-153.
Do I need to include a ribosomal binding site (RBS/Shine Dalgarno sequence) or Kozak sequence when I clone my gene of interest?
ATG is often sufficient for efficient translation initiation although it depends upon the gene of interest. The best advice is to keep the native start site found in the cDNA unless one knows that it is not functionally ideal. If concerned about expression, it is advisable to test two constructs, one with the native start site and the other with a Shine Dalgarno sequence/RBS or consensus Kozak sequence (ACCAUGG), as the case may be. In general, all expression vectors that have an N-terminal fusion will already have a RBS or initiation site for translation.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
Can you tell me the difference between a Shine-Dalgarno sequence and a Kozak sequence?
Prokaryotic mRNAs contain a Shine-Dalgarno sequence, also known as a ribosome binding site (RBS), which is composed of the polypurine sequence AGGAGG located just 5’ of the AUG initiation codon. This sequence allows the message to bind efficiently to the ribosome due to its complementarity with the 3’-end of the 16S rRNA. Similarly, eukaryotic (and specifically mammalian) mRNA also contains sequence information important for efficient translation. However, this sequence, termed a Kozak sequence, is not a true ribosome binding site, but rather a translation initiation enhancer. The Kozak consensus sequence is ACCAUGG, where AUG is the initiation codon. A purine (A/G) in position -3 has a dominant effect; with a pyrimidine (C/T) in position -3, translation becomes more sensitive to changes in positions -1, -2, and +4. Expression levels can be reduced up to 95% when the -3 position is changed from a purine to pyrimidine. The +4 position has less influence on expression levels where approximately 50% reduction is seen. See the following references:
- Kozak, M. (1986) Point mutations define a sequence flanking the AUG initiator codon that modulates translation by eukaryotic ribosomes. Cell 44, 283-292.
- Kozak, M. (1987) At least six nucleotides preceding the AUG initiator codon enhance translation in mammalian cells. J. Mol. Biol. 196, 947-950.
- Kozak, M. (1987) An analysis of 5´-noncoding sequences from 699 vertebrate messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
- Kozak, M. (1989) The scanning model for translation: An update. J. Cell Biol. 108, 229-241.
- Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res. 18, 2828.
Note: The optimal Kozak sequence for Drosophila differs slightly, and yeast do not follow this rule at all. See the following references:
- Romanos, M.A., Scorer, C.A., Clare, J.J. (1992) Foreign gene expression in yeast: a review. Yeast 8, 423-488.
- Cavaneer, D.R. (1987) Comparison of the consensus sequence flanking translational start sites in Drosophila and vertebrates. Nucleic Acids Res. 15, 1353-1361.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.