Chlorocyclohexane is used as the raw material in synthesis agrochemical and medicine chemical engineering products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorocyclohexane is used as the raw material in synthesis agrochemical and medicine chemical engineering products.
Solubility
Soluble in water.0.02 g/L (20°C). Soluble in alcohol and diethyl ether.
Notes
Incompatible materials are Strong oxidizing agents, Strong bases.
RUO – Research Use Only
Kenneth D. M. Harris and John M. Thomas. Structural aspects of the chlorocyclohexane/thiourea inclusion system.J. Chem. Soc., Faraday Trans.1990, 86 1095-1101.
Robert W. Crowe; Charles P. Smyth. The Dielectric and Polymorphic Behavior of Cyclohexanol, Cyclohexanone, Chlorocyclohexane and Cyclohexane.J. Am. Chem. Soc.1951, 73 (11), 5406-5411.