Thermo Scientific™

Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads, Thermo Scientific™

250g, Each
Thermo Scientific™

Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads, Thermo Scientific™

250g, Each
Quantity
Catalog number: L05466.30
also known as L05466-30
Price (USD)
38.00 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
70955-01-0
Appearance (Color)
Pale brown
Comment
Water absorbtion >20% @ 50%rH, 20?C, 24h
Residual water
≤1.5% w/w @ 550?C
Particle Size
≤5% is < 2.5mm
Form
Beads

Description

Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads are useful in general gas drying applications.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

RUO – Research Use Only

Figures