Dowex™ 1X8 200-400 (Cl) is used for fine chemical processing. It is used to determine trace levels of rhodium and platinum in environmetntal and biological samples. It is also used in anion-exchange chromatography for separation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dowex|r 1X8 200-400 (Cl) is used for fine chemical processing. It is used to determine trace levels of rhodium and platinum in environmetntal and biological samples. It is also used in anion-exchange chromatography for separation.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Strongly basic anion-exchange resin, 8% crosslinked, Cl- form, 200-400 mesh (dry).
Li, W. B.; Song, Y. B.; Xu, H. K.; Chen, L. Y.; Dai, W. H.; Dong, M. Ion-exchange method in the collection of nitrate from freshwater ecosystems for nitrogen and oxygen isotope analysis: a review. Environ. Sci. Pollut. Res. 2015, 22 (13), 9575-9588.
Königshofer, H.; Löppert, H. G. Regulation of invertase activity in different root zones of wheat (Triticum aestivum L.) seedlings in the course of osmotic adjustment under water deficit conditions. J. Plant Physiol. 2015, 183, 130-137.