Product Image
Invitrogen™

Invitrolon™ PVDF/Filter Paper Sandwiches, 0.45 μm, 8.5 x 13.5 cm

Invitrolon PVDF/Filter Paper Sandwiches, 0.45 μm, have excellent binding properties for western blotting, dot-blot assays, and protein sequencing. The 0.45-μmRead more
Have Questions?
Catalog NumberQuantity
LC200716 membrane/filter paper sandwiches
Catalog number LC2007
Price (USD)
452.65
Online exclusive
467.00
Save 14.35
Each
Quantity:
16 membrane/filter paper sandwiches
Price (USD)
452.65
Online exclusive
467.00
Save 14.35
Each
Invitrolon PVDF/Filter Paper Sandwiches, 0.45 μm, have excellent binding properties for western blotting, dot-blot assays, and protein sequencing. The 0.45-μm pore size makes these membranes ideal for protein transfer for proteins >10 kDa. The included extra-thick absorbent blotting paper (2.5 mm) accommodates blotting set-up using semi-dry transfer apparatuses.

See all membranes and filter papers ›

Features
• Membrane: polyvinylidene difluoride (PVDF)
• Filter paper thickness: 2.5 mm
• Binding capacity: goat IgG: 294 μg/cm2; BSA: 131 μg/cm2; insulin: 85 μg/cm2
• Re-probe characteristics: yes
• Pre-activation: required with 100% alcohol (methanol)
• Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, chemiluminescence, and radiolabeling
• Durability: compatible with most organic solvents, acids, and mild bases
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity16 membrane/filter paper sandwiches
Shipping ConditionRoom Temperature
Dimensions (LxW)8.5 cm x 13.5 cm
FormatSandwich
Length (Metric)13.5 cm
MaterialPVDF
Pore Size0.45 μm
Product LineInvitrolon Novex
Thickness2.5 mm
Width (Metric)8.5 cm
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

How can I store, strip, and reuse my western blot?

For nitrocellulose or PVDF membrane following Western blot detection using a chemiluminescent or fluorescent substrate system: Following transfer, air dry the membrane and place in an envelope, preferably on top of a supported surface to keep the membrane flat. The blot can be stored indefinitely at -80 degrees C. When ready to reprobe, prewet the PVDF blot with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with blocking step.

FOR STRIPPING/REPROBING OF MEMBRANES: Harsh protocol (see NOTE below for modifications)

1) Submerge the membrane in stripping buffer (100 mM BME, 2% SDS, 62.5 mM Tris-HCl, pH 6.7) and incubate at 50 degrees C for 30 min with occasional agitation. If more stringent conditions necessary, incubate at 70 degrees C.

2) Wash 2 x 10 min in TBS-T/PBS-T at room temperature.

3) Block the membrane by immersing in 5% blocking reagent TBS-T or PBS-T for 1 hr at room temperature.

4) Immunodetection

NOTE: Often you don't need such harsh conditions to remove antibodies from their proteins. The stringency of one or several of the variables can be decreased: lower the temperature, decrease the time, less BME, less SDS, etc. An especially mild but still often effective stripping protocol is lower pH incubation. Example: pH 2.0 Tris 50-100 mM, 30-60 min incubation (you may do two incubations if you wish). Then rinse and block as usual. If you do not wish to re-use the membrane immediately after stripping, you can store the membrane in plastic wrap (wet, you do not want it to dry out). Another simple, mild stripping buffer is 0.1 M glycine•HCl (pH 2.5-3.0), incubation 30 min to 2 hrs room temperature or 37 degrees C, depending on the antibody.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How do I destain proteins on a PVDF membrane that were stained with SimplyBlue SafeStain?

After staining with SimplyBlue SafeStain, use deionized water for the less strongly retained protein bands on the PVDF membrane.

Increasing methanol or ethanol concentrations up to 70% should destain any remaining bands. You can leave the membrane in the destain indefinitely.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I performed a western transfer onto a PVDF membrane and the transfer efficiency was very poor. Can you offer some tips?

Here are possible causes and solutions:

- The membrane may not be properly treated prior to transfer: Make sure that the membrane is pre-wetted with a polar organic solvent such as methanol or ethanol.
- There may be poor gel to membrane contact: Ensure that the filter paper and blotting pads are well saturated with transfer buffer, taking care to remove any bubbles during the assembly of the membrane sandwich. The gel/membrane sandwich must fit securely in the two halves of the blot module. Try adding another pad or replace any pads that have lost their resiliency with fresh ones.
- Over-compression of the gel: A good indication of over-compression is if the gel has been excessively flattened. In the event that the sandwich is over-compressed, remove enough pads so that the blotter can be closed without exerting excess pressure on the gel and the membrane.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Are your PVDF and nitrocellulose membranes compatible with the Li-COR instrument?

Yes, both our PVDF and nitrocellulose membranes are compatible with the Li-COR instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am planning on doing a dot blot and my sample contains acetonitrile. Can your PVDF or nitrocellulose membrane withstand acetonitrile?

Our PVDF can tolerate acetonitrile but our nitrocellulose cannot.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.