Thermo Scientific™

Lab Vision™ Vascular Endothelial Growth Factor (VEGF) Ab-7, Mouse Monoclonal Antibody

 Related applications:
Anatomical Pathology
Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-7, Mouse Monoclonal Antibody.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

VEGF which is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.

Host Species: Mouse

Clone: VG1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant VEGF protein

Molecular Weight: 19-22kDa (reduced)

Positive Control: VEGF165 recombinant protein, Angiosarcoma

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-7 does not neutralize the bioactivity of VEGF.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

General References:

1. Turley H, et al. (1998) J. Pathol. 186: 313-318.

2. Olyrne K J, et al. (2000) Br. J. Cancer. 82: 1427-1432.



Documents

Manuals & protocols