Attention: Product NI2400, StainEase™ Staining Trays will be discontinued November 2026 or while supplies last. See recommended alternative Catalog number: 5705-1010, Nalgene™ Staining Box or Catalog number: GSB-3, Owl™ Gel Staining Box. Please contact Technical Support with any questions.
Product Image
Invitrogen™
StainEase™ Staining Trays
The StainEase¤ Gel Staining Tray is a convenient way to evenly stain mini-gels and prevent gels from breaking. The stainingRead more
Have Questions?
Catalog NumberQuantity
NI24002 pack
Catalog number NI2400
Price (USD)
152.65
Online exclusive
157.00
Save 4.35
Each
Quantity:
2 pack
Request bulk or custom format
Price (USD)
152.65
Online exclusive
157.00
Save 4.35
Each
The StainEase¤ Gel Staining Tray is a convenient way to evenly stain mini-gels and prevent gels from breaking. The staining tray assembly includes a perforated strainer insert that holds the gel and drains away the stain, a lid, and an outer container in which stains and solutions can be changed without having to handle the gel. Easily fits up to four mini-gels.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity2 pack
Shipping ConditionRoom Temperature
Product LineStainEase
Product TypeStaining Tray
Unit SizeEach
Contents & Storage
Contents: StainEase™ Gel Staining Tray comes with two clear polycarbonate containers, two white polycarbonate strainers, two clear polycarbonate lids with handles and waterproof lid labels.
Frequently asked questions (FAQs)
Do you offer staining trays for staining protein gels?
We offer the following staining trays for staining protein gels:
- StainEase Staining Trays (Cat. No. NI2400) for convenient and even staining of Mini and Midi gels. The staining tray assembly includes a perforated strainer insert that holds the gel and drains away the stain, a lid, and an outer container in which stains and solutions can be changed without having to handle the gel. Each tray easily fits up to 4 mini gels.
- Mini Gel Incubation Trays (Cat. No. 22843) for staining mini gels and western blot membranes.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.