KT5720 PKA Inhibitor

Catalog number: PHZ1141

Invitrogen™  Related applications: Mammalian Cell Culture | Pharma & Biopharma | Protein Assays and Analysis | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

KT5720 is a potent, cell permeable inhibitor of PKA, and is a valuable tool in studies delineating cAMP-dependent signaling pathways, including activation of the transcription factor CREB. Inhibition of PKC, PKG, and MLCK is negligible. KT5720 is prepared by a chemical modification of K-252a.

Format/Formulation: White solid
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Solid
Inhibitor Properties: Chemical
Inhibitor Type: Kinase Inhibitor, Kinase Inhibitor (PKA Inhibitor)
Molecular Weight: 537.6 Da
Product Size: 0.1 mg
Purification: HPLC
Specific Inhibitor Target: PKA
Sterility: Nonsterile
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols