MK.1TE ESD and Static Latch-up Test System
MK.1TE ESD and Static Latch-up Test System
MK.1TE ESD and Static Latch-up Test System
Thermo Scientific™

MK.1TE ESD and Static Latch-up Test System

The Thermo Scientific™ MK.1TE is a relay-based, exceptionally fast ESD and Static Latch-Up Test System used in the evaluation of advanced IC devices. It fully addresses today's JEDEC/ESDA standards, and can be configured with 64, 128, 192 or 256 test pins.
Catalog NumberProduct Type
PIDESDMK1MK.1TE ESD and Static Latch-Up Test System
Catalog number PIDESDMK1
Price (GBP)
-
Product Type:
MK.1TE ESD and Static Latch-Up Test System

The Thermo Scientific™ MK.1TE ESD and Static Latch-up Test System provides users with advanced capabilities to test midrange pin count devices to today's Human Body Model (HBM) and Machine Model (MM) ESD standards. The system's pulse delivery design ensures recently identified waveform hazards in the standards, such as the trailing pulse and the pre-discharge voltage rise are addressed. Trailing pulses were shown to cause non-ESD related failures by exposing the DUT to an electrical overstress after the main HBM event. Pre-discharge voltage can cause voltage-triggered protection structures to fail, as the pin under test may not be at zero volts when the HBM event occurs. A user-selectable 10K Shunt can be connected during the pulse to eliminate any voltage prior to the actual HBM event. A combination test system, today's MK.1 test system also performs Static Latch-Up testing per JEDEC's EIA/JESD 78 method.

  • Waveform network: 8-site HBM pulse sources
  • Human Body Model (HBM) and Machine Model (MM) testing to industry standards
  • Static Latch-Up testing per current JEDEC’s EIA/JESD 78 method
  • Highly repeatable, reproducible test data
  • Enhanced dataset features
  • High voltage power supply chassis
  • Power supply sequencing
  • Event trigger output
  • Intuitive set-up and operation using the Thermo Scientific™ Scimitar™ software platform
  • Direct use of existing 256 pin-style ZapMaster test fixtures
Specifications
Dimensions (D x W x H)30.5 x 22.8 x 39 in. (77.5 x 57.9 x 99.1 cm)
Specifications Details

Enhanced Data Set Features:

Report all data gathered for off-line reduction and analysis; core test data is readily available; all data is stored in an easy-to-manipulate standard XML file structure. Scimitar™ operating software provides intuitive test setup and operation and enhanced data set features.

Product TypeMK.1TE ESD and Static Latch-Up Test System
Test Voltage Range30 V to 1 kV (MM); 30 V to 8 kV (HBM)
Voltage90/250 V
Unit SizeEach