Thermo Scientific™
MAS™ PTH Controls
 Related applications:
Diagnostic Testing
Accurately evaluate automated PTH methods with Thermo Scientific™ MAS™ PTH Controls. These ready-to-use, assayed, liquid controls are value assigned for key automated PTH methods and are conveniently packaged in a tri-level multi-pack.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Liquid, ready-to-use
  • Human serum based matrix
  • Assayed values for most major analyzers
Documents
Manuals & protocols