NDP Sensor Protein, phosphorylated

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant D112C mutant of Myxococcus xanthus nucleoside diphosphate kinase (NDPK) labeled with the fluorophore IDCC and phosphorylated on His117
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: Universal Kinase Assay
Detection Method: Fluorescent
Product Size: 10 µL
Target Enzyme: Kinases (Multiple)
Shipping Condition: Dry Ice

Contents & storage

Store in ultra-cold freezer (-68 to -85°C).

Documents

Manuals & protocols