LanthaScreen™ Tb-anti-c-fos [pThr232] Antibody

Catalog number:  PV5207

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

c-Fos is phosphorylated at Thr232 by mitogen-activated protein kinases (MAPK1 and MAPK3).
For use in combination with the LanthaScreen® c-Fos HeLa cell line (K1579) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Excitation⁄Emission (nm): 340⁄490, 520
Label or Dye: Tb (Terbium)
Product Line: LanthaScreen®
Product Size: 25 µg
Reactivity: Human
Readout: End Point
Target Gene: Ser/Thr Kinases
Shipping Condition: Dry Ice

Contents & storage

Terbium-labeled pc-fos (pSer73) antibody in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols