Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar (Emmons) w/Chloramphenicol. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concentration of dextrose from 40 to 20 g/l2. Chloramphenicol is added to the medium to make it inhibitory to bacterial overgrowth while allowing isolation of pathogenic fungi3.
Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol |
Format | Monoplate Deep Pour |
Product Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol |
Format | F - slant |
Product Type | Agar |
Quantity | 100/Pk. |
Unit Size | Each |
Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol |
Format | F - slant |
Product Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol |
Format | 1 oz. Bottle |
Product Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01770 | Each | 10/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08724 | Each | 100/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08726 | Each | 20/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R10158-MBD | Each | 10/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Agar (Emmons) w/Chloramphenicol.
This medium contains casein, meat peptones, nitrogenous compounds and amino acids necessary for the growth of fungi. Dextrose is an energy source which serves to support the growth of fungi. Chloramphenicol is a broad-spectrum antibiotic which inhibit a range of Gram-positive and Gram-negative bacteria. Yeast extract may be added to the medium as a growth supplement to ensure the growth of fastidious fungi.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.