Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons with Chloramphenicol is used for selective isolation of yeast and fungi.
Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol |
Format | 1 oz. Bottle |
Product Type | Agar |
Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol |
Format | Monoplate Deep Pour |
Product Type | Agar |
Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol |
Product Type | Agar |
Quantity | 100/Pk. |
Type | Agar |
Format | F - slant |
Unit Size | Each |
Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol |
Format | F - slant |
Product Type | Agar |
Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Format | Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R10158-MBD | Each | 1 oz. Bottle | 10/Pk. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R01770 | Each | Monoplate Deep Pour | 10/Pk. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08724 | Each | F - slant | 100/Pk. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08726 | Each | F - slant | 20/Pk. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|