Search Thermo Fisher Scientific
Cultivate anaerobic microorganisms using Cooked Meat Medium with Glucose, Hemin and Vitamin K.
Catalog Number | Quantity |
---|---|
R060542 | 20 x 7 mL |
R060540 | 100 x 7 mL |
Cultivate anaerobic microorganisms using Thermo Scientific™ Remel™ Cooked Meat Medium with Glucose, Hemin and Vitamin K. Robertson introduced the use of beef heart extract in water to support the growth of anaerobic bacilli1 while Henry used cooked beef heart to study saccharolytic and proteolytic properties of anaerobes2. This medium is also referred to as chopped meat medium. Cooked meat broth promotes growth of bacteria even from minute inocula and maintains viability of the culture for a long time.
Cultivate anaerobic microorganisms using Cooked Meat Medium with Glucose, Hemin and Vitamin K.
This liquid medium is recommended for use in qualitative procedures for cultivation of anaerobic microorganisms. It contains optimum salts, beef heart, peptone, and glucose which provide a reducing environment, amino acids and other nutrients necessary for the growth of anaerobes. Solid meat particles create favorable growth conditions for anaerobes due to reducing action of sulfhydryl group from muscle protein. Glucose provides energy for growth of microorganisms whereas Vitamin K and Hemin are essential for the growth of certain anerobes, especially Prevotella melaninogenica.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.