Thermo Scientific™

Remel™ Schaedler Broth

Catalog number: R064482
Thermo Scientific™

Remel™ Schaedler Broth

Catalog number: R064482

Thermo Scientific™ Remel Schaedler Broth is for the cultivation of fastidious aerobic and anaerobic microorganisms.

 
Catalog Number
R064482
Unit Size
Each
Quantity
20 x 5 mL
Price (USD)
Full specifications
Format15mm x 103mm Tube
TypeBroth
DescriptionSchaedler Broth
Product TypeBroth
Quantity20 x 5 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
100 x 5 mL
Price (USD)
Full specifications
Format15mm x 103mm Tube
TypeBroth
DescriptionSchaedler Broth
Product TypeBroth
Quantity100 x 5 mL
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
R064482Full specifications
Each20 x 5 mLRequest A Quote
Format15mm x 103mm Tube
TypeBroth
DescriptionSchaedler Broth
Product TypeBroth
Quantity20 x 5 mL
Unit SizeEach
R064480Full specifications
Each100 x 5 mLRequest A Quote
Format15mm x 103mm Tube
TypeBroth
DescriptionSchaedler Broth
Product TypeBroth
Quantity100 x 5 mL
Unit SizeEach
Showing 2 of 2

Thermo Scientific™ Remel Schaedler Broth is for the cultivation of fastidious aerobic and anaerobic microorganisms.

Figures