Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Cultivate a wide variety of microorganisms using Mueller Hinton Broth with Cations.
Catalog Number | Format | Packaging Type |
---|---|---|
R07127 | 16 x 125mm Tube - 10mL | 100/Pk. |
R07128 | 16 x 125mm Tube - 10mL | 20/Pk. |
R09598 | 20mm x 150mm Tube - 25mL | 20/Pk. |
Cultivate a wide variety of microorganisms using Thermo Scientific™ Remel™ Mueller Hinton Broth with Cations. Mueller Hinton formulated this medium as a protein-free medium for the purpose of isolating pathogenic Neisseria1. This was later used for testing gonococci and other organisms for susceptibility to sulfonamides2,3. More recently, MH Broth with Cations has become useful as a general-purpose cultivation broth.
Cultivate a wide variety of microorganisms with Mueller Hinton Broth with Cations.
Mueller Hinton Broth is prepared with beef extract and acid digest of casein that supply amino acids, nitrogenous substances, and other nutrients which are necessary for bacterial growth. Starch serves as a growth factor and a protective colloid that neutralizes toxic products which may be formed during the growth of bacteria. Calcium and Magnesium are divalent cations that enrich the medium.