Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Description | Format | Quantity | Manufacturing Origin |
---|---|---|---|---|
R09747 | Sabouraud Dextrose Agar, pH 5.6 | 20mm x 150mm Tube - Slant | 20/Pk. | United States |
R01766 | Sabouraud Dextrose Agar, pH 5.6 | Monoplate Deep Pour | 100/Pk. | United States |
R01768 | Sabouraud Dextrose Agar, pH 5.6 | Monoplate Deep Pour | 10/Pk. | United States |
R09746 | Sabouraud Dextrose Agar, pH 5.6 | 20mm x 150mm Tube - Slant | 100/Pk. | United States |
R112551 | Sabouraud Dextrose Agar, pH 5.6 | BB - 500mL | 10/Pk. | United States |
R112562 | Sabouraud Dextrose Agar, pH 5.6 | BRB - 100mL | 10/Pk. | United States |
R09744 | Sabouraud Dextrose Agar, pH 5.6 | 20 x 150mm Pour Tube (G-PT) | 100/Pk. | United States |