Search Thermo Fisher Scientific
Catalog Number | Format |
---|---|
R112417 | |
R110212 | |
R112418 | Borosilicate Bottle - 500mL |
Thermo Scientific™ Remel Letheen Agar is used for testing of quaternary ammonium compounds for antimicrobial activity.