Search Thermo Fisher Scientific
Thermo Scientific™ Remel Iodine for Tetrathionate is used with Tetrathionate Broth.
Description | Iodine for Tetrathionate Broth |
Format | Bottle |
Product Type | Iodine |
Type | Liquid |
Packaging Type | Bottle |
Quantity | 100 mL |
Unit Size | Each |
Description | Iodine for Tetrathionate Broth |
Format | Bottle |
Product Type | Iodine |
Type | Liquid |
Packaging Type | Bottle |
Quantity | 4.5 mL |
Unit Size | Each |
Description | Iodine for Tetrathionate Broth |
Format | Bottle |
Product Type | Iodine |
Type | Liquid |
Packaging Type | Bottle |
Quantity | 500 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R114350 | Full specifications | Each | 100 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R21228 | Full specifications | Each | 4.5 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R114351 | Full specifications | Each | 500 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel Iodine for Tetrathionate is used with Tetrathionate Broth.