This product is only available through your local Thermo Scientific supplier. Learn where to buy >
Wash Buffer 2 for GeneJET RNA Purification Kits (concentrated)
Catalog number: R1561
Thermo Scientific™  Related applications: RNA Extraction
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Thermo Scientific GeneJET Wash Buffer 2 (concentrated) is a component of the GeneJET RNA Purification Kit (K0731/K0732), the GeneJET Whole Blood RNA Purification Mini Kit (K0761) and the GeneJET Plant RNA Purification Mini Kit (K0801, K0802) and may be purchased separately. Provided in 100 mL aliquots.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Buffer Type: Wash Buffer
Product Line: GeneJET
Product Size: 100 mL
Documents
Manuals & protocols