Search Thermo Fisher Scientific
Rapidly detect and quantify specific antibodies to Brucella abortus and Brucella melitensis in sera with this stained suspension.
Catalog Number | Description | Detectable Analytes |
---|---|---|
R30855501 | Stained Brucella Suspensions | Brucella abortus |
R30953501 | Stained Brucella Suspensions | Brucella melitensis Antiodies |
Use Stained Brucella Suspensions for the qualitative detection of Brucella abortus and Brucella melitensis antibodies in sera in the investigation of pyrexia and in standard Widal tests1, 2, 3.
Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate bacterial suspensions which carry homologous antigens.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
1. Farrell, I.D., Robertson, L. et al. (1975). Serum antibody response in acute brucellosis. J. Hyg. (Camb.), 74, 23.
2. Kerr, W.R., McCaughey, W.J. et al. (1968). Techniques and interpretations in the serological diagnosis of Brucellosis in man. J. Med. Microbiol., 1, 181.
3. Quaife, R.A. (1969). Brucellosis in man. J. Med. Lab. Technol., 26, 349.