Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
R452674 | 2.5 kg |
R452672 | 500 g |
R452676 | 10 kg |
Thermo Scientific™ Remel Buffered Peptone Water is used for the non-selective pre-enrichment of Salmonella spp. from foods.