Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
R7310060 | Uni-OF Glucose |
R7310062 | Uni-OF Maltose |
R7310063 | Uni-OF Mannitol; |
R7310064 | Uni-OF Sucrose |
R7310065 | Uni-OF Xylose |
R7310061 | Uni-OF Lactose |
Remel Uni-OF facilitates determination of oxidative and fermentative carbohydrate metabolism of microorganisms.
Ordering Information:
Constricted tube does not require an oil overlay.