Thermo Scientific™

Lab Vision™ Wilm's Tumor Protein (WT1), Rabbit Polyclonal Antibody

Related applications:

Anatomical Pathology

Provide accurate, reproducible results with the Thermo Scientific™ Wilm's Tumor Protein (WT1), Rabbit Polyclonal Antibody.
Back to top
Wilm's Tumor (WT), a sporadic and familial childhood kidney tumor, is genetically heterogeneous. Wilm's tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed infetal kidney and mesothelioma, and its expression has been suggested as a marker for Wilm's tumor and mesothelioma.

Host Species: Rabbit

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of human Wilm's tumor protein

Molecular Weight: 47-55kDa

Positive Control: Wilm's tumor

Cellular Localization: Cytoplasmic and nuclear This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO