SYPRO™ Protein Gel Stains
Invitrogen™

SYPRO™ Protein Gel Stains

SYPRO protein gel stains are sensitive, easy-to-use fluorescent stains for total proteins separated by polyacrylamide gel electrophoresis (PAGE).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorQuantity
S12000Ruby1 L
S6650Orange500 μL
S6651Orange10 x 50 μL
S12010Tangerine500 μL
S6653Red500 μL
S6654
also known as S-6654
Red10 x 50 μL
S12001Ruby200 mL
S21900Ruby5 L
S12000X3Ruby3 x 1 L
Catalog number S12000
Price (USD)
463.65
Online exclusive
483.00
Save 19.35
Each
Color:
Ruby
Quantity:
1 L
Request bulk or custom format
Price (USD)
463.65
Online exclusive
483.00
Save 19.35
Each

SYPRO protein gel stains are sensitive, easy-to-use fluorescent stains for total proteins separated by polyacrylamide gel electrophoresis (PAGE). Stained proteins can be viewed with a standard UV or blue-light transilluminator, or imaging equipment containing the appropriate filters or laser. SYPRO protein gel stains offer many advantages over Coomassie Blue and silver staining, including a fast, one-step staining protocol requiring no destaining, a linear dynamic range over three orders of magnitude, and very little protein-to-protein variation in staining.

SYPRO Ruby Protein Gel Stain features include:

  • Convenient—ready-to-use formulation
  • High sensitivity—detection to 0.25 to 1 ng
  • Effective staining for 1D and 2D gels
  • Can be multiplexed with other gel stains such as Pro-Q Diamond phosphoprotein gel stain or Pro-Q Emerald 300 glycoprotein gel stain
  • Compatible with subsequent analysis of proteins by Edman-based sequencing or mass spectrometry

SYPRO Orange and SYPRO Red protein gel stain features include:

  • Sensitive—detection to 4 to 8 ng
  • Fast—rapid staining time of 10–60 min
  • Best for 1D gels
  • SYPRO Orange: Ex: 300, 470 nm, Em: 570 nm
  • SYPRO Red: Ex: 300, 550 nm, Em: 630 nm

SYPRO Tangerine Protein Gel Stain features include:

  • Sensitive—detection to 4 to 8 ng
  • Fast—rapid staining time of 10 to 60 minutes
  • Best for 1D gels
  • Does not require fixation with acetic acid
  • Compatible with subsequent gel staining, western blotting, zymography, electroelution or mass spectrometry
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1X
Detection LocationIn-Gel Detection
Detection MethodFluorescence
Excitation/Emission280, 450/610 nm
Quantity1 L
Shelf Life9 Months
Shipping ConditionRoom Temperature
Target MoleculeProtein
ColorRuby
Label or DyeSYPRO Ruby
Product LineSYPRO
Product TypeProtein Gel Stain
Unit SizeEach
Contents & Storage
Store at room temperature and protect from light.

Frequently asked questions (FAQs)

Can proteins from 1D SDS-PAGE gels be transferred to PVDF or nitrocellulose after being stained with SYPRO Ruby protein gel stain?

No. Proteins stained with SYPRO Ruby protein gel stain cannot be blotted onto membranes. The fixation step and the fixative-like solution that the dye is dispersed in prevents efficient blotting of proteins onto membranes.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am getting a broad 50-68 kDa band across the entire length of the gel when I stain with SYPRO Ruby Protein Gel Stain and other protein stains. What is this and how can I prevent it?

Your samples or the gel wells were contaminated with keratins from skin or hair. Rinse out the gel wells with ultrapure water or running buffer before loading samples. Wear a lab coat and gloves when preparing samples and use microfuge tubes that have been stored in sealed plastic bags, not left out on the bench top, for preparing samples.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Some of my pre-stained molecular weight markers are appearing as dark bands when I stain the gel with SYPRO Ruby Protein Gel Stain. What is causing this?

Blue-colored dyes absorb light in the red wavelengths, so they absorb the red fluorescent emission of SYPRO Ruby dye. SYPRO Ruby dye still binds these proteins, but the signal is quenched by the colored dye, resulting in a negatively stained, dark band. Examples of molecular weight markers with blue-colored proteins that will quench SYPRO Ruby fluorescence are the BenchMark Pre-Stained Protein Ladder and some proteins in the SeeBlue Plus2 Pre-Stained Standard. The same phenomenon can be seen with the bromophenol blue dye front, if it is not completely run off the gel, and loss of signal when SYPRO Ruby stained gels are subsequently stained with Coomassie Blue stains. Most other colored dyes do not quench the SYPRO Ruby dye signal and will appear as normally stained protein bands.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

My gels are showing a large amount of ‘speckles' after staining with SYPRO Ruby Protein Gel Stain. How are they formed and what can I do to remove them or prevent them from even forming?

Speckles on the gel can increase as the SYPRO Ruby Protein Gel Stain ages, due to self-aggregation of the SYPRO Ruby dye over time. Speckles can also form due to dye aggregation around contaminants from the staining container, solutions, or particles from the air or gloves, including keratin proteins from skin and hair. When gels are incubated with SYPRO Ruby Protein Gel Stain for several hours or longer, dye can build up on the sides of the staining container and then be dislodged with continuing rocking, especially during the destain step, forming speckles. Non-dye speckles can also show up in the image from auto-fluorescent particles of dust, hair, glove powder, or clothing lint that falls on the gel or surface of the glass imaging plate. The better the imager is at focusing on surface features of the gel, the more speckles that are going to be visible.

To minimize the formation of speckles and other background debris, follow clean laboratory practices, use ultrapure water of greater than 18 megohm-cm resistance to prepare solutions, rinse gloves in water to remove powder residue before touching gels, use lint-free wipes and wear a lab coat or avoid wearing clothing that generates a lot of lint, always rinse the staining container with ethanol and wipe out any residual dye before staining another gel, and always rinse and wipe down the glass imaging surface with ethanol and water before placing your gel down. Remove dye buildup on the surface of the staining dish by wiping out the dish with ethanol between the stain and wash step. The rapid stain protocol is complete in as little as 90 minutes, which does not allow enough time for most speckles for form. Once speckles have been deposited on the gel, it is not possible to wash them off. Speckles will show up as sharp, tall spikes on 3D renditions of gel images. These spikes look distinct from 3D renditions of protein spots or bands. Some image analysis software packages have de-speckling algorithms that can easily identify and remove this type of pixelated noise.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

My SYPRO Ruby Protein Gel Stain is more than a year old and has some visible precipitate. Can I filter the stain to remove the precipitate and reduce the amount of ‘speckles' on the surface of my stained gels?

SYPRO Ruby Protein Gel Stain is not stable beyond about a year. The dye begins to precipitate out from solution (self-aggregate) over time and will show a lower staining intensity of protein bands and increased ‘debris' or ‘speckles' on the surface of the gel. It is not possible to filter the stain to remove dye precipitate, as the dye sticks to most paper and membrane filters and will be removed from the staining solution.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (254)

Citations & References
Abstract
The major CD9 and CD81 molecular partner. Identification and characterization of the complexes.
Authors:Charrin S,Le Naour F,Oualid M,Billard M,Faure G,Hanash SM,Boucheix C,Rubinstein E
Journal:The Journal of biological chemistry
PubMed ID:11278880
Autotaxin is released from adipocytes, catalyzes lysophosphatidic acid synthesis, and activates preadipocyte proliferation. Up-regulated expression with adipocyte differentiation and obesity.
Authors:Ferry G,Tellier E,Try A,Grés S,Naime I,Simon MF,Rodriguez M,Boucher J,Tack I,Gesta S,Chomarat P,Dieu M,Raes M,Galizzi JP,Valet P,Boutin JA,Saulnier-Blache JS
Journal:The Journal of biological chemistry
PubMed ID:12642576
Our group has recently demonstrated (Gesta, S., Simon, M., Rey, A., Sibrac, D., Girard, A., Lafontan, M., Valet, P., and Saulnier-Blache, J. S. (2002) J. Lipid Res. 43, 904-910) the presence, in adipocyte conditioned-medium, of a soluble lysophospholipase d-activity (LPLDact) involved in synthesis of the bioactive phospholipid lysophosphatidic acid (LPA). ... More
Changes in global gene and protein expression during early mouse liver carcinogenesis induced by non-genotoxic model carcinogens oxazepam and Wyeth-14,643.
Authors:Iida M,Anna CH,Hartis J,Bruno M,Wetmore B,Dubin JR,Sieber S,Bennett L,Cunningham ML,Paules RS,Tomer KB,Houle CD,Merrick AB,Sills RC,Devereux TR
Journal:Carcinogenesis
PubMed ID:12727805
We hypothesized that the mouse liver tumor response to non-genotoxic carcinogens would involve some common early gene and protein expression changes that could ultimately be used to predict chemical hepatocarcinogenesis. In order to identify a panel of genes to test, we analyzed global differences in gene and protein expression in ... More
Correlation between checkpoint activation and in vivo assembly of the yeast checkpoint complex Rad17-Mec3-Ddc1.
Authors:Giannattasio M, Sabbioneda S, Minuzzo M, Plevani P, Muzi-Falconi M
Journal:J Biol Chem
PubMed ID:12672803
Rad17-Mec3-Ddc1 forms a proliferating cell nuclear antigen-like complex that is required for the DNA damage response in Saccharomyces cerevisiae and acts at an early step of the signal transduction cascade activated by DNA lesions. We used the mec3-dn allele, which causes a dominant negative checkpoint defect in G1 but not ... More
Short-term training enhances endothelium-dependent dilation of coronary arteries, not arterioles.
Authors:Laughlin MH, Rubin LJ, Rush JW, Price EM, Schrage WG, Woodman CR
Journal:J Appl Physiol
PubMed ID:12391095
Our objective was to test the hypothesis that short-term exercise training (STR) of pigs increases endothelium-dependent dilation (EDD) of coronary arteries but not coronary arterioles. Female Yucatan miniature swine ran on a treadmill for 1 h, at 3.5 mph, twice daily for 7 days (STR; n = 28). Skeletal muscle ... More