SYBR Safe™ DNA gel stain was developed specifically for reduced mutagenicity—to be safer than ethidium bromide for staining DNA inRead more
Have Questions?
Change view
Catalog Number
Quantity
S33111
1 L
S33112
4 L
2 Options
Catalog number S33111
Price (USD)
160.65
Online exclusive
171.00
Save 10.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
160.65
Online exclusive
171.00
Save 10.35
Each
SYBR Safe™ DNA gel stain was developed specifically for reduced mutagenicity—to be safer than ethidium bromide for staining DNA in agarose or acrylamide gels. SYBR Safe™stain is not only less mutagenic than ethidium bromide, but SYBR Safe™ stain's detection sensitivity is better than that of ethidium bromide. SYBR Safe™ stain comes as a premixed solution that can be used just like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe™ stain has fluorescence excitation maxima at ∼280 and ∼502 nm, and an emission maximum at ∼530 nm.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection LocationIn-Gel Detection
Detection MethodFluorescence
Quantity1 L
Shelf Life6 months from date of purchase
Shipping ConditionRoom Temperature
Target MoleculeDNA
Label or DyeSYBR Safe
Product TypeDNA Gel Stain
Unit SizeEach
Contents & Storage
• Supplied as a ready-to-use solution in 1X TAE
Store at room temperature in the original container.
Frequently asked questions (FAQs)
Why do I sometimes see speckles in my gel when using SYBR Safe DNA Gel Stain?
Many whitening agents used in clothing, as well as some fungi and bacteria, fluoresce at the same wavelengths as SYBR Safe DNA gel stain. These contaminants within or on the surface of the gel may produce this speckling.
What is the pH range of SYBR dyes?
The SYBR dyes are useful only over a narrow range of pH, from about 7 to 8. Outside this range, the fluorescent signal diminishes rapidly.
Which direction does the SYBR Safe dye run during electrophoresis?
Similarly to ethidium bromide, SYBR Safe DNA Gel Stain runs in the opposite direction of the migrating DNA. This has no practical effect on the use of gels cast with SYBR Safe DNA Gel Stain, as only the very bottom of the gel will have a lower concentration of stain. This effect can be partially counteracted by staining the gel with SYBR Safe DNA Gel Stain after electrophoresis. Solutions of dye should not be added to the running buffer as this can cause breakdown of the dye at the electrodes and release toxic volatile compounds into the air.
Does ethanol precipitation remove the SYBR Safe dye?
SYBR Safe DNA Gel Stain is easily removed from nucleic acids by ethanol precipitation.
Can I reuse SYBR Safe DNA Gel Stain for a second gel?
We strongly discourage the reuse of SYBR Safe DNA Gel Stain, as this practice significantly lowers sensitivity.
Mechanisms of palatal epithelial seam disintegration by transforming growth factor (TGF) beta3.
Authors:Ahmed S, Liu CC, Nawshad A,
Journal:Dev Biol
PubMed ID:17698055
TGFbeta3 signaling initiates and completes sequential phases of cellular differentiation that is required for complete disintegration of the palatal medial edge seam, that progresses between 14 and 17 embryonic days in the murine system, which is necessary in establishing confluence of the palatal stroma. Understanding the cellular mechanism of palatal ... More