SabIR™ Diffuse Reflectance Probe
Thermo Scientific™

SabIR™ Diffuse Reflectance Probe

Thermo Scientific SabIR Diffuse Reflectance Probe is a high-performance NIR fiber optic accessory for the Antaris™ FT-NIR analyzer.
Have Questions?
Catalog NumberFor Use With (Equipment)Sample TypeType
SABIRDIFFUSEThermo Scientific Antaris II and MX FT-NIR spectrometersSolids and PowdersSabIR Diffuse Reflectance Probe
Catalog number SABIRDIFFUSE
Price (HKD)
-
For Use With (Equipment):
Thermo Scientific Antaris II and MX FT-NIR spectrometers
Sample Type:
Solids and Powders
Type:
SabIR Diffuse Reflectance Probe
Thermo Scientific SabIR Diffuse Reflectance Probe is a high-performance NIR fiber optic accessory for the Antaris™ FT-NIR analyzer. Ideal for quality control laboratories that need remote sampling capabilities, the SabIR Diffuse Reflectance Probe provides excellent analysis of pharmaceuticals, polymers, fabrics, food and biological samples.
  • Enables remote non-destructive sampling through a variety of packaging materials
  • Particularly useful for raw material identification, quality measurements, and sample component analysis
  • Remote analysis of powders and solids
  • Stainless steel construction and rugged sapphire window
  • Remote “start” trigger with status lights
  • Two, three and five meter lengths available
  • Standard SMA fiber optic probe connection
  • Integrated Spectralon reference standard
With the Transflectance Adapter, the SabIR probe can be used to analyze liquid samples.
Specifications
DescriptionThermo Scientific SabIR Diffuse Reflectance Probe is a high-performance NIR fiber optic accessory for the Antaris FT-NIR analyzer.
Sample TypeSolids and Powders
For Use With (Equipment)Thermo Scientific Antaris II and MX FT-NIR spectrometers
TypeSabIR Diffuse Reflectance Probe
Unit SizeEach