Impeller sleeve for open-top mixing, for 500/1000L HyPerforma™ Single-Use Mixer BioProcess Container (BPC)
Thermo Scientific™
Impeller sleeve for open-top mixing, for 500/1000L HyPerforma™ Single-Use Mixer BioProcess Container (BPC)
The Impeller sleeve for open-top mixing (500/1000L) supports mixing applications in bioproduction using Thermo Scientific™ Bioprocess Containers (BPCs) for ThermoRead more
Have Questions?
Catalog number SH30749.10
Price (USD)
-
Request A Quote
The Impeller sleeve for open-top mixing (500/1000L) supports mixing applications in bioproduction using Thermo Scientific™ Bioprocess Containers (BPCs) for Thermo Scientific™ HyPerforma™ Single-Use Mixer, with impeller-based mixing in open or closed designs.
Single Use Mixer (S.U.M.) BPC options include: • Open top liner for media/buffer hydration • Closed BPC with powder port for contained media/buffer hydration • Closed BPC with powder port and monitoring probe capability • Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions • Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
For Use With (Equipment)HyPerforma 1000 L Single-Use Mixer, HyPerforma 500 L Single-Use Mixer