Save valuable lab space with Thermo Scientific™ MaxQ™ 6000 Incubated/Refrigerated Stackable Shakers, which offer sample storage and orbital shaking with intuitive, easy-to-use digital controls. These shakers save space by stacking units two-high on the floor. Use the adapter set (available separately) to tier two 18 x 18″ platforms to double shaking capacity.
Two adjustable-height shelves provide added storage
Choose from two temperature ranges to fulfill a variety of molecular biology, protein expression or plasmid purification studies
Available in incubated or refrigerated models
Precisely monitor and control chamber temperature over complete range with ±0.1°C accuracy at 37°C with PID temperature controller
Overtemperature safety feature with independent thermostat provides additional backup by controlling heat if main temperature controller fails
Solid-state DC brushless motor, variable speed control from 15 to 500rpm
Continuous and timed operation from 0.1 hour up to 999 hours or 0.1 minute to 999 minutes
View speed, operating time and temperature simultaneously on three individual LED displays, which help minimize errors in establishing running conditions
Displayed time counts up during continuous operation and down during timed runs
Visual/audible signals alert user to temperature deviations; heat turns off if temperature deviates ±1°C of setpoint
Shaker shutdown and audible/visual alarms signal if unit operates ±10% of set speed, preventing the shaker from walking
Eliminate sudden starts and stops with soft start feature
User-adjustable speed calibration can be performed using a digital handheld tachometer, where verification is required and protocols can be standardized
Safety interlock stops shaking motion when the door is open
Platform motion stops when excess vibration is detected due to an unbalanced load
Retains parameters during power failure and restarts unit automatically after power is restored
RS-232 interface allows remote monitoring of speed, time and temperature
Large viewing window and internal light allow sample visibility, minimizing the need to open the door
Door can be hinged from the right or left by the user for convenient placement in the lab
18 × 18 in. platform can be dual-tiered inside the chamber to double the shaking capacity
Corrosion-resistant stainless-steel chamber accommodates a variety of 18 × 18 in. (46 × 46cm) universal and dedicated platforms
Durable powder-coated, cold-rolled steel exterior construction
Electrical outlet inside chamber provides power for safe operation of shakers, stirrers or rotators
Overall Dimensions
Platform dimensions: 18 × 18 in. (45.7 × 45.7cm)
Overal dimensions L × W × H: 33 × 27 × 40.5 in. (83.9 × 69.6 × 102.9cm)
Ordering Information:
For a complete system, order platforms and clamps (sold separately)
Includes:
Two adjustable-height perforated stainless-steel shelves.