The 50 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (2 plug + pinch clamp, 1 injection site).
Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions
Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands
BPC port specifications:
• Line 1: Plug 6.4 mm (0.25 in.), pinch clamp. Tubing: C-Flex 374, length: 51 cm (20 in.), ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
• Line 2: Plug 6.4 mm (0.25 in.), pinch clamp. Tubing: C-Flex 374, length: 51 cm (20 in.), ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
• Line 3: Luer injection port. Tubing: C-Flex, length: 7.6 cm (3 in.)