Limited time offer: 3 year warranty when you purchase a SimpliAmp Thermal Cycler. View flyer »

PikoReal 24, validation plate (1 plate)

Catalog number: TCR0024VAL

Thermo Scientific™  Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific PikoReal 24 Validation Plate is designed to verify the performance of the Thermo Scientific PikoReal 24 RealTime PCR System.

The Validation Plate is a pre-loaded and sealed 24-well plate. Each well contains 20 µl of reaction mixture with the same quantity of Lambda DNA template in each reaction. The qPCR mixture of the PikoReal 24 Validation Plate is green to visualize the reaction mixture in each well and evaluate equal volumes in each well before and after cycling.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): PikoReal™ Real-Time PCR System
Product Line: PikoReal™
Product Size: 1 each
Type: qPCR plate

Contents & storage

Contains:
• 1 × 24-well Validation plate Store at -20°C.

Documents

Manuals & protocols