Search Thermo Fisher Scientific
Certifications/Compliance | CE Marked |
Description | Infinity Carbon Dioxide |
CE Marker | Yes |
Detectable Analytes | Carbon Dioxide |
Format | Liquid |
Sample Type | Serum |
Type | Reagent |
Linearity | 3 to 50mEq/L |
Methods Compatibility | Enzymatic Endpoint |
Quantity | 2 x 125 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TR28321 | Full specifications | Each | 2 x 125 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|