Eliminate incorrect card insertion with the polarized Thermo Scientific™ Filter-Holders for Neutron Monitoring.
Have Questions?
Catalog number TYPE8806
Price (USD)/ Each
-
Request A Quote
Description:
TYPE885X
Eliminate incorrect card insertion with the polarized Thermo Scientific™ Filter-Holders for Neutron Monitoring. They are gasket-sealed to exclude dirt and moisture.
Equipped with tamper-evident seals
Provided with visual indication of card barcode ID through window
Color-coded per customer specification, barcoded if so specified
TYPE885X Filter holder is compatible with Type 8806B (strap clip and belt loops attachment), 8806S (strap clip and strap loop attachment), and 8816 (strap clip and strap loop attachment).
Specifications
Product TypeFilter Holder
CompatibilityType 8806B (strap clip and belt loops attachment), 8806S (strap clip and strap loop attachment), and 8816 (strap clip and strap loop attachment).